ContactUsLinkComponent
NANOCOLOR ECO Ammonium LR tube test measuring range: 0,01 - 1,80 mg/L NH4-N 0,01 - 2,30 mg/L NH4+ sufficient for 100 determinations 1 * 1 KIT
Codice catalogo MANA976003
Nome del fornitore:  MACHEREY-NAGEL
NANOCOLOR ECO Ammonium LR tube test measuring range: 0,01 - 1,80 mg/L NH4-N 0,01 - 2,30 mg/L NH4+ sufficient for 100 determinations 1 * 1 KIT
Codice catalogo MANA976003
Nome del fornitore:  MACHEREY-NAGEL
Frequently Bought Together