ContactUsLinkComponent
I-191 IS A POTENT, SELECTIVE PROTEASE-ACTIVATED RECEPTOR 2 (PAR2) ANTAGONIST THAT INHIBITS MULTIPLE PAR2-INDUCED SIGNALING PATHWAYS AND FUNCTIONAL RESPONSES. 1 * 5 mg
Catalog # SELKE0102-5MG
Supplier:  SELLECK CHEMICALS
I-191 IS A POTENT, SELECTIVE PROTEASE-ACTIVATED RECEPTOR 2 (PAR2) ANTAGONIST THAT INHIBITS MULTIPLE PAR2-INDUCED SIGNALING PATHWAYS AND FUNCTIONAL RESPONSES. 1 * 5 mg
Catalog # SELKE0102-5MG
Supplier:  SELLECK CHEMICALS
Frequently Bought Together