ContactUsLinkComponent
SARS-CoV-2 S protein RBD (K417T,E484K,N501Y), Fc Tag (SPD-C5258) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T, E484K, N501Y)). The K417T, E484K, N501Y mutations were identified in the SA 1 * 1 mg
Catalog # ACRBSPD-C5258-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 S protein RBD (K417T,E484K,N501Y), Fc Tag (SPD-C5258) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T, E484K, N501Y)). The K417T, E484K, N501Y mutations were identified in the SA 1 * 1 mg
Catalog # ACRBSPD-C5258-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together