ContactUsLinkComponent
SARS-CoV-2 Nucleocapsid protein (P80R), His Tag (NUN-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 4 19 (Accession # QHO62115.1 (P80R)). The nucleocapsid protein is consisted of the N-terminal RNA-binding domain (NTD) 1 * 100 µG
Catalog # ACRBNUN-C52HC100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Nucleocapsid protein (P80R), His Tag (NUN-C52Hc) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 4 19 (Accession # QHO62115.1 (P80R)). The nucleocapsid protein is consisted of the N-terminal RNA-binding domain (NTD) 1 * 100 µG
Catalog # ACRBNUN-C52HC100UG
Supplier:  ACRO Biosystems
Frequently Bought Together