SARS-CoV-2 Spike RBD (L452R, E484Q), His Tag (SPD-C52Hv) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession # QHD43416.1 (L452R, E484Q)). The L452R, E484Q mutations were identified in the SARS-CoV-2 variants whic 1 * 1 mg
Catalog # ACRBSPD-C52HV-1MG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together