HCoV-229E Spike Protein, His Tag (SPN-H52H3) is expressed from human 293 cells (HEK293). It contains AA Cys 16 - Pro 1115 (Accession # P15423-1 (TI 871-872 PP)).The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trim 1 * 50 µG
Catalog # ACRBSPN-H52H3-50UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together