ContactUsLinkComponent
PE-Labeled Human PSMA, His Tag (PSA-HP2Q3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1). 1 * 25 Tests
Catalog # ACRBPSAHP2Q325TEST
Supplier:  ACRO Biosystems
PE-Labeled Human PSMA, His Tag (PSA-HP2Q3) is expressed from human 293 cells (HEK293). It contains AA Lys 44 - Ala 750 (Accession # Q04609-1). 1 * 25 Tests
Catalog # ACRBPSAHP2Q325TEST
Supplier:  ACRO Biosystems
Frequently Bought Together