ContactUsLinkComponent
Mouse IL-2, His Tag (IL2-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Gln 169 (Accession # P04351-1). 1 * 100 µG
Catalog # ACRBIL2-M52H3100UG
Supplier:  ACRO Biosystems
Mouse IL-2, His Tag (IL2-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Gln 169 (Accession # P04351-1). 1 * 100 µG
Catalog # ACRBIL2-M52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together