ContactUsLinkComponent
Biotinylated Cynomolgus IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag (ILG-C82F6) is expressed from human 293 cells (HEK293). It contain s AA Ala 27 - Thr 240 (IL-2RB) & Glu 22 - Glu 238 (IL-2RA) & Leu 23 - Pro 255 (IL-2RG) (Accession # Q38J85-1 (IL-2RB 1 * 200 µG
Catalog # ACRBILG-C82F6200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Cynomolgus IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag (ILG-C82F6) is expressed from human 293 cells (HEK293). It contain s AA Ala 27 - Thr 240 (IL-2RB) & Glu 22 - Glu 238 (IL-2RA) & Leu 23 - Pro 255 (IL-2RG) (Accession # Q38J85-1 (IL-2RB 1 * 200 µG
Catalog # ACRBILG-C82F6200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together