ContactUsLinkComponent
PE-Labeled Human Her2, His Tag (HE2-HP2E3) is produced via conjugation of PE to Human Her2, His Tag under optimal conditions. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). 1 * 25 Tests
Catalog # ACRBHE2HP2E325TEST
Supplier:  ACRO Biosystems
PE-Labeled Human Her2, His Tag (HE2-HP2E3) is produced via conjugation of PE to Human Her2, His Tag under optimal conditions. Human Her2, His Tag is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession # P04626-1). 1 * 25 Tests
Catalog # ACRBHE2HP2E325TEST
Supplier:  ACRO Biosystems
Frequently Bought Together