ContactUsLinkComponent
Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1). 1 * 20 µG
Catalog # ACRBCD9-M52H3-20UG
Supplier:  ACRO Biosystems
Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1). 1 * 20 µG
Catalog # ACRBCD9-M52H3-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together