ContactUsLinkComponent
Biotinylated Human IL-11, His,Avitag (IL1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Access ion # P20809-1). 1 * 200 µG
Catalog # ACRBIL1-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-11, His,Avitag (IL1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Leu 199 (Access ion # P20809-1). 1 * 200 µG
Catalog # ACRBIL1-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together