ContactUsLinkComponent
Mouse IL-15 R alpha, Fc Tag (ILA-M5253) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - Lys 205 (Accession # Q60 819-1). 1 * 100 µG
Catalog # ACRBILA-M5253100UG
Supplier:  ACRO Biosystems
Mouse IL-15 R alpha, Fc Tag (ILA-M5253) is expressed from human 293 cells (HEK293). It contains AA Gly 33 - Lys 205 (Accession # Q60 819-1). 1 * 100 µG
Catalog # ACRBILA-M5253100UG
Supplier:  ACRO Biosystems
Frequently Bought Together