ContactUsLinkComponent
Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1). 1 * 100 µG
Catalog # ACRBBAR-R52H3100UG
Supplier:  ACROBIOSYSTEMS
Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1). 1 * 100 µG
Catalog # ACRBBAR-R52H3100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together