ContactUsLinkComponent
Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1). 1 * 1 mg
Codice catalogo ACRBBAR-R52H3-1MG
Nome del fornitore:  ACROBIOSYSTEMS
Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1). 1 * 1 mg
Codice catalogo ACRBBAR-R52H3-1MG
Nome del fornitore:  ACROBIOSYSTEMS
Frequently Bought Together