ContactUsLinkComponent
QuickSeal qPCR Optical Film (100 sheets).A transparent, optically clear, DMSO resistant, non-tacky film which adheres only when pressure is applied. It is non-pierceable and peelable. Clear plastic, reflective, glossy on the top. Very thin and light 1 * 100 items
Catalog # ISTSIST-128-080LS
Supplier:  iST Scientific Ltd
QuickSeal qPCR Optical Film (100 sheets).A transparent, optically clear, DMSO resistant, non-tacky film which adheres only when pressure is applied. It is non-pierceable and peelable. Clear plastic, reflective, glossy on the top. Very thin and light 1 * 100 items
Catalog # ISTSIST-128-080LS
Supplier:  iST Scientific Ltd
Frequently Bought Together