ContactUsLinkComponent
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ht) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion QHD43416.1). Mutations T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681R, D950N are id 1 * 50 µG
Catalog # ACRBSPN-C52HT-50UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Ht) is the ectodomain of SARS-CoV-2 spike protein which contains AA Val 16 - Pro 1213 (Acce ssion QHD43416.1). Mutations T19R, V70F, FR157-158Del, A222V, W258L, K417N, L452R, T478K, D614G, P681R, D950N are id 1 * 50 µG
Catalog # ACRBSPN-C52HT-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together