ContactUsLinkComponent
SARS-CoV-2 Spike RBD (K417N, L452R, T478K), His Tag (SPD-C5226) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession QHD43416.1 (K417N, L452R, T478K)). The mutations K417N, L452R, T478K are identified in the SARS- 1 * 1 mg
Catalog # ACRBSPD-C5226-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD (K417N, L452R, T478K), His Tag (SPD-C5226) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession QHD43416.1 (K417N, L452R, T478K)). The mutations K417N, L452R, T478K are identified in the SARS- 1 * 1 mg
Catalog # ACRBSPD-C5226-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together