ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (SPD-C82Eh) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 5 37 (Accession QHD43416.1 (L452Q, F490S)). Mutations L452Q, F490S are identified in the SARS-CoV-2 Lambda Variant (C. 1 * 25 µG
Catalog # ACRBSPD-C82EH-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike RBD, His,Avitag (SPD-C82Eh) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 5 37 (Accession QHD43416.1 (L452Q, F490S)). Mutations L452Q, F490S are identified in the SARS-CoV-2 Lambda Variant (C. 1 * 25 µG
Catalog # ACRBSPD-C82EH-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together