Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Papain

Papain

Poly bottle, 500 g
Catalog # SPCMPA125-500GM
Supplier:   Spectrum Chemical
Papain
Catalog # SPCMPA125-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Papain

Poly bottle, 500 g

CAS Number:  
Price
SFr. 462.00
Log in for availability
1 * 500 g
Product Details & Documents

Papain, powder, purified, is a cysteine protease enzyme the is present in the papaya fruit.

It is traditionally used to break down meat fibres. It has more recently been used to dissociate cells in the early steps of cell culture preparation.

Documents

Specifications

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Cat. No.:
    SPCMPA125-500GM
  • Form:
    Powder, purified
  • Purity:
    Purified
  • Enzyme name:
    Papain

Specifications