Apcin, Chemical. CAS: 300815-04-7. Formula: C13H14Cl3N7O4. MW: 438.7. Cell permeable compound capable of slowing anaphase-promoting complex/cyclosome APC/C-dependent proteolysis. Inhibitor of the ubiquitin ligase activity of the APC/C by inhibiting i 1 * 25 mg
Catalog # ADIPAGCR13603M025
Supplier: AdipoGen Life Sciences
Frequently Bought Together