Celite® Standard Super-cel, filter aid, GPR RECTAPUR®
HDPE bucket, 5 kg
Supplier: VWR Chemicals
Celite® Standard Super-cel, filter aid, GPR RECTAPUR®
Catalog # 22556.360
Supplier: VWR Chemicals
CAS Number: 91053-39-3
Jump to:
Product Details & Documents
Celite® is used as a filtering aid or as adsorbent in column chromatography.
- Filter aid
- Calcined, fresh water diatomeous earth
- CAS Number:91053-39-3
- MDL Number:MFCD00132803
Documents
Certificates
Search by Lot Numberspec_VWRC22556.290 spec_VWRC22556.360
Spécification du produit
VWR® for Drying and Filtration
Documentation produit
Specifications
Specification Test Results
- pH (25 °C; 10% in water ):8,5 - 11,0
- Wet density:≤ 357 g/l
- Loss on ignition (1000 °C):≤ 0.5%
- Particle size (< 56µm):≥ 90%
Specifications
- Pack type:HDPE bucket
- Pk:5 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



