ContactUsLinkComponent
Tip Cell Saver 1-200µl Natural Loose Non-Sterile with Wide Bore Tip to Prevent Shearing when Pipetting Cells 1 * 1.000 items
Catalog # ALPCLW1018
Supplier:  Alpha Laboratories
Tip Cell Saver 1-200µl Natural Loose Non-Sterile with Wide Bore Tip to Prevent Shearing when Pipetting Cells 1 * 1.000 items
Catalog # ALPCLW1018
Supplier:  Alpha Laboratories
Frequently Bought Together