Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Digitonin

Bottle, 250 mg
Catalog # APOSBID3301-250MG
Toxic
Supplier:   Apollo Scientific
Digitonin
Catalog # APOSBID3301-250MG
Supplier:  Apollo Scientific
CAS Number:  11024-24-1
Digitonin

Bottle, 250 mg

CAS Number:  11024-24-1
Price
SFr. 65.90
Log in for availability
1 * 250 mg
Product Details & Documents

Non-ionic detergent used to permeabilize cell membranes.

Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • Storage Temperature:Ambient
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications