ContactUsLinkComponent
Binocular ERGO brightfield microscope, 1000x, IOS W-PLAN objectives 1 * 1 items
Catalog # OPTKB-510ERGO
Supplier:  OPTIKA
Binocular ERGO brightfield microscope, 1000x, IOS W-PLAN objectives 1 * 1 items
Catalog # OPTKB-510ERGO
Supplier:  OPTIKA
Frequently Bought Together