Sodium thiosulfate solution c(Na2S2O3) = 0.05 mol/l (0.05 N) Titripur 1 * 1 L
Catalog # 1.60311.1003
Supplier: Merck
Sodium thiosulfate solution c(Na2S2O3) = 0.05 mol/l (0.05 N) Titripur 1 * 1 L
Catalog # 1.60311.1003
Supplier: Merck
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Zinc, granules, EMSURE® ISO analytical reagent, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals