ContactUsLinkComponent
Smooth stainless steel sheet, folded and rebated at the front and laterally by 1.6'' (40 mm), rear side with 1.6'' (40 mm) upstand and rebated. Welded/polished corners. Shelf load: 30.2 lbs/ft (60kg/m) 1 * 1 items
Catalog # HUPF0163428
Supplier:  HUPFER METALLWERKE
Smooth stainless steel sheet, folded and rebated at the front and laterally by 1.6'' (40 mm), rear side with 1.6'' (40 mm) upstand and rebated. Welded/polished corners. Shelf load: 30.2 lbs/ft (60kg/m) 1 * 1 items
Catalog # HUPF0163428
Supplier:  HUPFER METALLWERKE
Frequently Bought Together