ContactUsLinkComponent
ClearView Blackfly S Colour 0.4 MP USB3 Vision Camera, Sony IMX287 CMOS, 1/2.9 ,6.9 µm, Global shutter, 720 x 540 at 522 FPS, CS-mou nt. 1 * 1 ST
Catalog # MSPPBFSU304S2CCS
ClearView Blackfly S Colour 0.4 MP USB3 Vision Camera, Sony IMX287 CMOS, 1/2.9 ,6.9 µm, Global shutter, 720 x 540 at 522 FPS, CS-mou nt. 1 * 1 ST
Catalog # MSPPBFSU304S2CCS
Frequently Bought Together