10-100um Polydisperse Standards 10 bottle set 0.10g 1 * 0,1 g
Catalog # WHITPS312-0.10
Supplier: WHITEHOUSE
10-100um Polydisperse Standards 10 bottle set 0.10g 1 * 0,1 g
Catalog # WHITPS312-0.10
Supplier: WHITEHOUSE
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Supplier: VWR Chemicals
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
Sodium borohydride analytical reagent, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
Sieve Standards, Polydisperse Particle Standard
Supplier: WHITEHOUSE
pH buffer, single-use, SINGLET™
Supplier: Hach