Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Fall Arrest Roofing Kits

Fall Arrest Roofing Kits

Fall arrest roofing kit
Catalog # PRTWFP66RER
Supplier:   Portwest
Fall Arrest Roofing Kits
Catalog # PRTWFP66RER
Supplier:  Portwest
Fall Arrest Roofing Kits
Fall Arrest Roofing Kits

Fall arrest roofing kit

Price
SFr. 227.00
Log in for availability
1 * 1 KIT
Product Details & Documents

Kit includes two-point harness, carabiner, 12 mm detachable rope grab, 10 m kernmantle static rope, and nylon drawstring bag.

  • Chest loops
  • High-quality 12 mm kernmantle rope
  • Maximum rated load: 100 kg

The versatile drawstring bag is high-strength yet lightweight, making it ideal for securely holding fall protection products.

10-year shelf life from manufacturing date outlined on product label.

Specifications
  • Description:
    Fall arrest roofing kit
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.