Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Vessel dilators, balanced instruments

Vessel dilators, balanced instruments

Stainless steel, 150 mm, Suture tying platform, Straight, Ø 0,3 mm
Catalog # FSCI18602-15
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vessel dilators, balanced instruments
Catalog # FSCI18602-15
Supplier:  FINE SCIENCE TOOLS
Marketsource
Vessel dilators, balanced instruments
Vessel dilators, balanced instruments

Stainless steel, 150 mm, Suture tying platform, Straight, Ø 0,3 mm

Price
SFr. 252.84
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    150 mm
  • Material:
    Stainless steel
  • Seq:
    03
  • Tip style:
    Suture tying platform
  • Tip size:
    Ø 0,3 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.