Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Vessel cannulation forceps

Vessel cannulation forceps

0,6 mm Ext.Ø tubing size, Inox, 135 mm, Angled
Catalog # FSCI18406-13
Marketsource
Supplier:   FINE SCIENCE TOOLS
Vessel cannulation forceps
Catalog # FSCI18406-13
Supplier:  FINE SCIENCE TOOLS
Marketsource
Vessel cannulation forceps
Vessel cannulation forceps

0,6 mm Ext.Ø tubing size, Inox, 135 mm, Angled

Price
116.80 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    0,6 mm Ext.Ø tubing size
  • Length:
    135 mm
  • Material:
    Inox
  • Seq:
    03
  • Tip shape:
    Angled