ContactUsLinkComponent
HygroLab including probe HC2-AW, Sample holder: WP-40, Disposable sample container PS-14 (100 pc.) + PS-40(100 pc.) 1 * 1 SET
Catalog # ROTRHYGROLABSET40
HygroLab including probe HC2-AW, Sample holder: WP-40, Disposable sample container PS-14 (100 pc.) + PS-40(100 pc.) 1 * 1 SET
Catalog # ROTRHYGROLABSET40
Frequently Bought Together