Order Entry
Switzerland
Orders LinkContactUsLinkComponent
2',3'-Dimethoxyacetophenone 95%

2',3'-Dimethoxyacetophenone 95%

Bottle, 1 g
Catalog # APOSOR938335-1G
Supplier:   Apollo Scientific
2',3'-Dimethoxyacetophenone 95%
Catalog # APOSOR938335-1G
Supplier:  Apollo Scientific
CAS Number:  38480-94-3
2',3'-Dimethoxyacetophenone 95%

Bottle, 1 g

CAS Number:  38480-94-3
Price
36.40 SFr.
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:38480-94-3
  • Formula:C₁₀H₁₂O₃
  • MDL Number:MFCD03424412
  • Molecular Weight:180.2005 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.