Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tridecanoic acid 99%

Tridecanoic acid 99%

Bottle, 5 g
Catalog # APOSOR54705-5G
Irritant
Supplier:   Apollo Scientific
Tridecanoic acid 99%
Catalog # APOSOR54705-5G
Supplier:  Apollo Scientific
CAS Number:  638-53-9
Tridecanoic acid 99%

Bottle, 5 g

CAS Number:  638-53-9
Price
SFr. 26.00
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00002741
  • Molecular Weight:214.35 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.