Tridecanoic acid 99%
Bottle, 25 g
Katalognr. APOSOR54705-25G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: n-Tridecanoic acid, Tridecylic acid
- CAS Nummer:638-53-9
- Formel:C₁₃H₂₆O₂
- MDL-Nummer:MFCD00002741
- Molekulargewicht:214.35 g/mol
- Lagertemperatur:Freezer
Technische Daten
Technische Daten
- Pack type:Bottle
- Pk:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



