Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Tridecanoic acid 99%

Tridecanoic acid 99%

Bottle, 100 g
Catalog # APOSOR54705-100G
Irritant
Supplier:   Apollo Scientific
Tridecanoic acid 99%
Catalog # APOSOR54705-100G
Supplier:  Apollo Scientific
CAS Number:  638-53-9
Tridecanoic acid 99%

Bottle, 100 g

CAS Number:  638-53-9
Price
SFr. 326.00
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  n-Tridecanoic acid, Tridecylic acid
  • CAS Number:638-53-9
  • Formula:C₁₃H₂₆O₂
  • MDL Number:MFCD00002741
  • Molecular Weight:214.35 g/mol
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications