Order Entry
Switzerland
Orders LinkContactUsLinkComponent
D(-)-Isoleucine

D(-)-Isoleucine

Bottle, 250 mg
Catalog # APOSOR36341-250MG
Supplier:   Apollo Scientific
D(-)-Isoleucine
Catalog # APOSOR36341-250MG
Supplier:  Apollo Scientific
CAS Number:  319-78-8
D(-)-Isoleucine

Bottle, 250 mg

CAS Number:  319-78-8
Price
91.80 SFr.
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  (2R,3R)-(-)-Isoleucine, H-D-Ile-OH
  • CAS Number:319-78-8
  • Formula:H₃CCH₂CH(CH₃)CH(NH₂)COOH
  • MDL Number:MFCD00064221
  • Molecular Weight:131.17 g/mol
  • Melting Pt:265…274 °C
  •  :Density: 1.224 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications