Order Entry
Switzerland
Orders LinkContactUsLinkComponent
O411 98+%

O411 98+%

Bottle, 250 mg
Catalog # APOSOR21534-250MG
Supplier:   Apollo Scientific
O411 98+%
Catalog # APOSOR21534-250MG
Supplier:  Apollo Scientific
CAS Number:  175135-47-4
O411 98+%

Bottle, 250 mg

CAS Number:  175135-47-4
Price
SFr. 369.00
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  4-[(4-Methoxyphenyl)methoxy]benzeneacetonitrile, 2-(4-((4-Methoxybenzyl)oxy)phenyl)acetonitrile, O4I1
  • CAS Number:175135-47-4
  • Formula:C₁₆H₁₅NO₂
  • MDL Number:MFCD00067778
  • Molecular Weight:253.30 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.