Titriplex IIIsolution c(Na2-EDTA) =0.05 mol/l (0.05N) (20 GRAD C) Titripur 1 * 1 L
Catalog # 1.60320.1003
Supplier: Merck
Titriplex IIIsolution c(Na2-EDTA) =0.05 mol/l (0.05N) (20 GRAD C) Titripur 1 * 1 L
Catalog # 1.60320.1003
Supplier: Merck
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Measuring Beakers with Handle, PMP TPX
Supplier: VWR Collection
Nitric acid 68%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Microscope slides
Supplier: MARIENFELD
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium thiosulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylenol orange tetrasodium salt, TECHNICAL
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection