Titriplex III solution c(Na2-EDTA) = 0.01 mol/l (0.01 N) (20 GRAD C) Titripur 1 * 1 L
Catalog # 1.60319.1003
Supplier: Merck
Titriplex III solution c(Na2-EDTA) = 0.01 mol/l (0.01 N) (20 GRAD C) Titripur 1 * 1 L
Catalog # 1.60319.1003
Supplier: Merck
Frequently Bought Together
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Nessler reagent A, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for Titrette®
Supplier: Brand
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Sodium thiosulphate 0.01 mol/l (0.01 N) in aqueous solution, VOLUSOL® volumetric solution
Supplier: VWR Chemicals
Nitric acid 65%, EMPLURA®, Supelco®
Supplier: Merck
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
VWR®, Nitrile Gloves
Supplier: VWR Collection
Ammonia - Ammonium Chloride Buffer Solution, pH 10
Supplier: CPACHEM