Propionic acid ≥99.7%, GPR RECTAPUR®
Plastic jerrican, 25 L
Supplier: VWR Chemicals
Propionic acid ≥99.7%, GPR RECTAPUR®
Catalog # 87062.460
Supplier: VWR Chemicals
CAS Number: 79-09-4
Jump to:
Product Details & Documents
Synonyms: Propanoic acid
- CAS Number:79-09-4
- Formula:C₃H₆O₂
- MDL Number:MFCD00002756
- Molecular Weight:74.08 g/mol
- Boiling Pt:140…142 °C
- Flash Pt:54 °C
- Storage Temperature:Ambient
- UN:3463
- ADR:8,II
Specifications
Specification Test Results
- Assay (GC) :min 99.0%
- Identity (IR) :passes test
- Density (d 20°C/4°C):0,993 – 0,994
- Water (KF):max 0.30%
- Appearance:clear liquid
Specifications
- Pack type:Plastic jerrican
- Pk:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



