ContactUsLinkComponent
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike RBD protein to streptavidin conjugated magnetic beads, whi ch can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The beads are in uniform size 1 * 2 mg
Catalog # ACRBMBS-K002-2MG
Supplier:  ACROBIOSYSTEMS
The pre-coupled magnetic beads coupled with biotinylated SARS-CoV-2 Spike RBD protein to streptavidin conjugated magnetic beads, whi ch can capture the Anti- SARS-CoV-2 antibody or ACE2 protein from cell or serum sample. The beads are in uniform size 1 * 2 mg
Catalog # ACRBMBS-K002-2MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together