ContactUsLinkComponent
Human CD7, His Tag CD7-H52H7 is expressed from human 293 cells HEK293. It contains AA Ala 26 - Pro 180 Accession P09564-1. 1 * 100 µG
Catalog # ACRBCD7-H52H7100UG
Supplier:  ACRO Biosystems
Human CD7, His Tag CD7-H52H7 is expressed from human 293 cells HEK293. It contains AA Ala 26 - Pro 180 Accession P09564-1. 1 * 100 µG
Catalog # ACRBCD7-H52H7100UG
Supplier:  ACRO Biosystems
Frequently Bought Together