ContactUsLinkComponent
Biotinylated Human CD20 Full Length, His,Avitag HEK293 CD0-H82E5 is expressed from human 293 cells HEK293. It contains AA Met 1 - Pr o 297 Accession P11836-1. 1 * 20 µG
Catalog # ACRBCD0-H82E5-20UG
Supplier:  ACRO Biosystems
Biotinylated Human CD20 Full Length, His,Avitag HEK293 CD0-H82E5 is expressed from human 293 cells HEK293. It contains AA Met 1 - Pr o 297 Accession P11836-1. 1 * 20 µG
Catalog # ACRBCD0-H82E5-20UG
Supplier:  ACRO Biosystems
Frequently Bought Together