ContactUsLinkComponent
Rhesus macaque CD32a, His Tag SPR  BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 50 µG
Catalog # ACRBCDA-R52H4-50UG
Supplier:  ACRO Biosystems
Rhesus macaque CD32a, His Tag SPR  BLI verified CDA-R52H4 is expressed from human 293 cells HEK293. It contains AA Gln 34 - Ile 217 Accession NP_001244229.1. 1 * 50 µG
Catalog # ACRBCDA-R52H4-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together