ContactUsLinkComponent
The biotinylated BCMA protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-BCMA antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
Catalog # ACRBMBS-K004-10MG
Supplier:  ACRO Biosystems
The biotinylated BCMA protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-BCMA antibody from various assay systems. The beads are in uniform size, narrow size distribution with large surf 1 * 10 mg
Catalog # ACRBMBS-K004-10MG
Supplier:  ACRO Biosystems
Frequently Bought Together