ContactUsLinkComponent
SARS-CoV-2 S protein RBD, Fc Tag SPD-C5255 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accession QHD 43416.1. 1 * 1 mg
Catalog # ACRBSPD-C5255-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD, Fc Tag SPD-C5255 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Phe 541 Accession QHD 43416.1. 1 * 1 mg
Catalog # ACRBSPD-C5255-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together