ContactUsLinkComponent
Biotinylated Human ACE2, His,Avitag AC2-H82E6 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q 9BYF1-1. 1 * 25 µG
Catalog # ACRBAC2-H82E6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human ACE2, His,Avitag AC2-H82E6 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q 9BYF1-1. 1 * 25 µG
Catalog # ACRBAC2-H82E6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together