ContactUsLinkComponent
Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag NUN-C81Q6 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Ac cession QHO62115.1. 1 * 200 µG
Catalog # ACRBNUN-C81Q6200UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Nucleocapsid protein, His,Avitag NUN-C81Q6 is expressed from E.coli cells. It contains AA Met 1 - Ala 419 Ac cession QHO62115.1. 1 * 200 µG
Catalog # ACRBNUN-C81Q6200UG
Supplier:  ACRO Biosystems
Frequently Bought Together